ENA336956511 Enamine

1-[2-(trifluoromethyl)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD12828681
  • Cas Number : 39959-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86035
Availability details for
Quantity
  • Availability
  • Add to Cart