ENA336956514 Enamine

ethyl({[3-(pyridin-4-yl)phenyl]methyl})amine

  • MDL number MFCD12785862
  • Cas Number : 1250627-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86043
Availability details for
Quantity
  • Availability
  • Add to Cart