ENA336956519 Enamine

1-benzylazetidine-2-carboxylic acid

  • MDL number MFCD04038976
  • Cas Number : 18085-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86104
Availability details for
Quantity
  • Availability
  • Add to Cart