ENA336956520 Enamine

(2S)-2-amino-3-(1H-indol-3-yl)-N-methylpropanamide

  • MDL number MFCD14590225
  • Cas Number : 53708-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84212
Availability details for
Quantity
  • Availability
  • Add to Cart