ENA341290534 Enamine

1-(chloromethyl)-4-(difluoromethane)sulfonylbenzene

  • MDL number MFCD20501886
  • Cas Number : 1354949-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63...65
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-49431

Safety Information

msds https://www.enaminestore.com/catalog/EN300-49431
Availability details for
Quantity
  • Availability
  • Add to Cart