ENA341290540 Enamine

5-(5-bromo-2-chlorophenyl)-1H-pyrazol-3-amine

  • MDL number MFCD12168578
  • Cas Number : 1186639-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64733

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64733
Availability details for
Quantity
  • Availability
  • Add to Cart