ENA341290544 Enamine

1-(2-ethylpiperazin-1-yl)propan-2-ol

  • MDL number MFCD20501897
  • Cas Number : 1354949-73-7
  • Molecular Weight : 172.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70949
Availability details for
Quantity
  • Availability
  • Add to Cart