ENA341290567 Enamine

tert-butyl N-({2,4-dioxo-1,3-diazaspiro[4.5]decan-6-yl}methyl)carbamate

  • MDL number MFCD20501919
  • Cas Number : 1354950-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85855
Availability details for
Quantity
  • Availability
  • Add to Cart