ENA341290570 Enamine

(2-bromoethyl)dibutylamine hydrobromide

  • MDL number MFCD00494132
  • Cas Number : 854392-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86604
Availability details for
Quantity
  • Availability
  • Add to Cart