ENA341290588 Enamine

2-bromo-1-(2,3-dihydro-1H-indol-1-yl)-3-methylbutan-1-one

  • MDL number MFCD09940757
  • Cas Number : 1016838-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29150
Availability details for
Quantity
  • Availability
  • Add to Cart