ENA341290589 Enamine

4-(2-bromo-3-methylbutanoyl)-1,2,3,4-tetrahydroquinoxalin-2-one

  • MDL number MFCD11538677
  • Cas Number : 1042811-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29156
Availability details for
Quantity
  • Availability
  • Add to Cart