ENA341290591 Enamine

tert-butyl N-{6-aminospiro[3.3]heptan-2-yl}carbamate

  • MDL number MFCD09054707
  • Cas Number : 1239589-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-119
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29519
Availability details for
Quantity
  • Availability
  • Add to Cart