ENA341290598 Enamine

N-[2-(2-fluorophenyl)ethyl]cyclopentanamine

  • MDL number MFCD11143328
  • Cas Number : 1019577-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32840

Safety Information

msds https://www.enaminestore.com/catalog/EN300-32840
Availability details for
Quantity
  • Availability
  • Add to Cart