ENA341290599 Enamine

1-(2,5-dichlorophenyl)ethan-1-amine

  • MDL number MFCD05215237
  • Cas Number : 603945-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33165
Availability details for
Quantity
  • Availability
  • Add to Cart