ENA341290605 Enamine

3-[2-(5-methylfuran-2-yl)ethenyl]-1H-1,2,4-triazole-5-thiol

  • Cas Number : 929971-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-34772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34772
Availability details for
Quantity
  • Availability
  • Add to Cart