ENA341290611 Enamine

3-(2-phenylethenyl)-1H-1,2,4-triazole-5-thiol

  • Cas Number : 218438-53-0
  • Molecular Weight : 203.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35141
Availability details for
Quantity
  • Availability
  • Add to Cart