ENA341290627 Enamine

3,9-diazabicyclo[3.3.2]decan-10-one

  • Cas Number : 97278-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39003
Availability details for
Quantity
  • Availability
  • Add to Cart