ENA341290645 Enamine

5-(1-chloroethyl)-3-(4-fluoro-3-methylphenyl)-1,2,4-oxadiazole

  • MDL number MFCD10694406
  • Cas Number : 1152532-51-8
  • Molecular Weight : 240.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-42474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42474
Availability details for
Quantity
  • Availability
  • Add to Cart