ENA341290717 Enamine

(1R,5S,6R)-3-[(tert-butoxy)carbonyl]-3-azabicyclo[3.1.0]hexane-6-carboxylic acid

  • MDL number MFCD12198680
  • Cas Number : 927679-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55188

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-55188
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart