ENA341290724 Enamine

2-[(tert-butoxy)carbonyl]-1,2,3,4-tetrahydroisoquinoline-1-carboxylic acid

  • MDL number MFCD08274995
  • Cas Number : 166591-85-1
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56243
Availability details for
Quantity
  • Availability
  • Add to Cart