ENA341290800 Enamine

2,2-dimethylpiperazine

  • MDL number MFCD06798288
  • Cas Number : 84477-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-35
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59819
Availability details for
Quantity
  • Availability
  • Add to Cart