ENA341290801 Enamine

methyl 2-[(piperidin-4-yl)methyl]furan-3-carboxylate hydrochloride

  • MDL number MFCD09878305
  • Cas Number : 1134618-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59960
Availability details for
Quantity
  • Availability
  • Add to Cart