ENA341290828 Enamine

3-(4,5-dimethyl-1,3-thiazol-2-yl)piperidine

  • MDL number MFCD09891525
  • Cas Number : 1017153-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62608

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62608
Availability details for
Quantity
  • Availability
  • Add to Cart