ENA341290837 Enamine

3-chloropyridazine

  • MDL number MFCD06801356
  • Cas Number : 1120-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-63951
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart