ENA341290844 Enamine

4,4-difluoro-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD01862059
  • Cas Number : 537033-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64292
Availability details for
Quantity
  • Availability
  • Add to Cart