ENA341290888 Enamine

N-methyl-1-(6-methylpyridazin-3-yl)piperidin-3-amine

  • MDL number MFCD16789072
  • Cas Number : 1247729-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70891

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70891
Availability details for
Quantity
  • Availability
  • Add to Cart