ENA341290898 Enamine

1-[(5-cyclopropyl-1,2,4-oxadiazol-3-yl)methyl]-1,4-diazepane

  • MDL number MFCD14679527
  • Cas Number : 1249732-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71070

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71070
Availability details for
Quantity
  • Availability
  • Add to Cart