ENA341290899 Enamine

2-[(1H-1,2,4-triazol-1-yl)methyl]piperidine

  • MDL number MFCD16075577
  • Cas Number : 1249074-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71160
Availability details for
Quantity
  • Availability
  • Add to Cart