ENA341290917 Enamine

2-methyl-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD00278415
  • Cas Number : 4165-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73092
Availability details for
Quantity
  • Availability
  • Add to Cart