ENA341290918 Enamine

2-azaspiro[3.3]heptane-6-carboxylic acid hydrochloride

  • MDL number MFCD11858156
  • Cas Number : 1172252-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73242

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73242
Availability details for
Quantity
  • Availability
  • Add to Cart