ENA341290927 Enamine

7-methyl-1,4-diazepan-5-one

  • MDL number MFCD19216491
  • Cas Number : 90673-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74029
Availability details for
Quantity
  • Availability
  • Add to Cart