ENA341290933 Enamine

4-chloroquinoline-2-carbonitrile

  • MDL number MFCD08705675
  • Cas Number : 4552-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74962

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74962
Availability details for
Quantity
  • Availability
  • Add to Cart