ENA341290948 Enamine

[1,2,4]triazolo[4,3-a]pyridine-6-carbonitrile

  • MDL number MFCD09994414
  • Cas Number : 1019024-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76322
Availability details for
Quantity
  • Availability
  • Add to Cart