ENA341290953 Enamine

7-[(tert-butoxy)carbonyl]-7-azabicyclo[2.2.1]heptane-2-carboxylic acid

  • MDL number MFCD18433537
  • Cas Number : 1221818-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77234
Availability details for
Quantity
  • Availability
  • Add to Cart