ENA341290955 Enamine

[1-(difluoromethyl)-1H-imidazol-2-yl]methanamine

  • MDL number MFCD13358357
  • Cas Number : 1094415-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24494
Availability details for
Quantity
  • Availability
  • Add to Cart