ENA341290979 Enamine

4-ethylmorpholine-2-carbonitrile

  • MDL number MFCD20441511
  • Cas Number : 1354950-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78699

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78699
Availability details for
Quantity
  • Availability
  • Add to Cart