ENA341290988 Enamine

4-(2-fluorophenyl)-2-methylbutanoic acid

  • MDL number MFCD16304268
  • Cas Number : 1225852-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80103

Safety Information

msds https://www.enaminestore.com/catalog/EN300-80103
Availability details for
Quantity
  • Availability
  • Add to Cart