ENA341290993 Enamine

4-[4-(difluoromethoxy)phenyl]oxan-4-amine

  • MDL number MFCD11186114
  • Cas Number : 1094442-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-80462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-80462
Availability details for
Quantity
  • Availability
  • Add to Cart