ENA341290996 Enamine

1-[(1,2-oxazol-3-yl)methyl]piperazine

  • MDL number MFCD06738163
  • Cas Number : 885952-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-81550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-81550
Availability details for
Quantity
  • Availability
  • Add to Cart