ENA341291002 Enamine

3-amino-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD01528030
  • Cas Number : 124004-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245-246
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-82138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82138
Availability details for
Quantity
  • Availability
  • Add to Cart