ENA341291003 Enamine

4-[4-(benzyloxy)phenyl]oxane-4-carboxylic acid

  • MDL number MFCD20441514
  • Cas Number : 1354953-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82144
Availability details for
Quantity
  • Availability
  • Add to Cart