ENA341291005 Enamine

4-bromobenzene-1-carboximidamide hydrochloride

  • MDL number MFCD00464967
  • Cas Number : 55368-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 262...264
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82342

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-82342
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart