ENA341291008 Enamine

2-{4-hydrazinyl-5H,6H,7H-cyclopenta[d]pyrimidin-2-yl}pyridine

  • MDL number MFCD17046871
  • Cas Number : 1343885-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82472
Availability details for
Quantity
  • Availability
  • Add to Cart