ENA341291009 Enamine

tert-butyl 2-methylpiperazine-1-carboxylate

  • MDL number MFCD02179168
  • Cas Number : 120737-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82748

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-82748
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart