ENA341291031 Enamine

2-[5-(acetamidomethyl)thiophene-2-sulfonamido]acetic acid

  • MDL number MFCD20441518
  • Cas Number : 1354963-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83903
Availability details for
Quantity
  • Availability
  • Add to Cart