ENA341291043 Enamine

(1R)-1-(4-fluoro-2-methoxyphenyl)ethan-1-ol

  • MDL number MFCD18340616
  • Cas Number : 1344936-21-5
  • Molecular Weight : 170.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-84205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84205
Availability details for
Quantity
  • Availability
  • Add to Cart