ENA341291053 Enamine

2-hydroxycyclopentane-1-carboxylic acid

  • MDL number MFCD17270607
  • Cas Number : 81887-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-84646
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart