ENA341291056 Enamine

2-(2,5-dihydro-1H-pyrrol-1-yl)ethan-1-amine

  • MDL number MFCD19216348
  • Cas Number : 34971-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84783

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84783
Availability details for
Quantity
  • Availability
  • Add to Cart