ENA341291063 Enamine

5-cyclopropyl-N-methyl-1H-pyrazol-3-amine

  • MDL number MFCD20441527
  • Cas Number : 1354949-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84890
Availability details for
Quantity
  • Availability
  • Add to Cart