ENA341291064 Enamine

methyl 4-amino-2,6-difluorobenzoate

  • MDL number MFCD16038807
  • Cas Number : 191478-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84899

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84899
Availability details for
Quantity
  • Availability
  • Add to Cart